Calcium Disodium EDTA, FCC, 97-102%, Spectrum™ Chemical
E1194, 23411-34-9, C10H12CaN2Na2O8•xH2O
Supplier: Spectrum Chemical Mfg Cor E1194500GM
Description
Spectrum™ Chemical Calcium Disodium EDTA, FCC is used as a flavor and color preservative. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
23411-34-9 | |
6.5-7.5 | |
C10H16CaN2Na2O9 | |
O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O | |
97 to 102% | |
Poly Bottle |
100% | |
500 g | |
WJEKUTCBXSHIOX-UHFFFAOYSA-J | |
394.30 | |
FCC |
Chemical Identifiers
23411-34-9 | |
394.30 | |
O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O |
C10H16CaN2Na2O9 | |
WJEKUTCBXSHIOX-UHFFFAOYSA-J |