missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium DL-glycerate dihydrate, Thermo Scientific Chemicals
$151.40 - $556.99
Chemical Identifiers
CAS | 67525-74-0 |
---|---|
Molecular Formula | C3H5O4 |
Molecular Weight (g/mol) | 105.07 |
MDL Number | MFCD00149289,MFCD00149288,MFCD00149289 |
InChI Key | RBNPOMFGQQGHHO-UWTATZPHSA-M |
Synonym | calciumdl-glycerate |
PubChem CID | 131883203 |
IUPAC Name | calcium;2,3-dihydroxypropanoic acid;dihydrate |
SMILES | OC[C@@H](O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0821403
|
Thermo Scientific Chemicals
L0821403 |
1 g |
Each for $151.40
|
|
|||||
AAL0821406
|
Thermo Scientific Chemicals
L0821406 |
5 g |
Each for $556.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in cold and hot water.
Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
67525-74-0 | |
105.07 | |
RBNPOMFGQQGHHO-UWTATZPHSA-M | |
131883203 | |
OC[C@@H](O)C([O-])=O |
C3H5O4 | |
MFCD00149289,MFCD00149288,MFCD00149289 | |
calciumdl-glycerate | |
calcium;2,3-dihydroxypropanoic acid;dihydrate |
Specifications
∽140°C (decomposition) | |
1 g | |
MFCD00149289,MFCD00149288,MFCD00149289 | |
14,4483 | |
Soluble in cold and hot water. | |
OC[C@@H](O)C([O-])=O | |
105.07 | |
286.26 (250.23 Anhydrous) |
67525-74-0 | |
C3H5O4 | |
4614104 | |
calciumdl-glycerate | |
RBNPOMFGQQGHHO-UWTATZPHSA-M | |
calcium;2,3-dihydroxypropanoic acid;dihydrate | |
131883203 | |
Calcium DL-glycerate dihydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 282-247-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only