Learn More
Calgonate Corp CALCIUM GLUCONATE SAFETY KIT DFS Item
Supplier:  Calgonate KITFIRSTAID01
This convenient emergency kit contains over 30 essential items for HF acid exposure response and general workplace first aid. It also contains 2 Tubes of Calgonate Gel (25g each) and 2 Bottles of Saline Solution (120ml each)
Catalog No. NC0200462
Only null left
Explore available promotions