missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium hydrogen phosphate, anhydrous, 98% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04023230

 View more versions of this product

Catalog No. AA4023230

Explore available promotions

Description

Description

It is used as a feed additive for livestock and poultry. It can promote feed digestion, put on animal’s weight, raise laying rate or producing milk rate, cure diseases such as rickets, osteomalacia and anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium hydrogen phosphate
7757-93-9
Powder
CaHO4P
MFCD00010909
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium;hydrogen phosphate
24441
136.06
Odorless
Anhydrous
White
250 g
CaHPO4
14,1692
Soluble in hydrochloric acid,nitric acid,acetic acid,ammonium citrate solution. Slightly soluble in water. Insoluble in ethanol.
[Ca++].OP([O-])([O-])=O
136.06
CHEBI:32596
≤98%

Chemical Identifiers

7757-93-9
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
24441
calcium;hydrogen phosphate
CaHO4P
MFCD00010909
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
CHEBI:32596
[Ca++].OP([O-])([O-])=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-826-1

RTECSNumber : TB8528000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only