Learn More
Calcium Hydroxide (Powder/Certified), Fisher Chemical™
Supplier: Fisher Chemical FLC97500
Specifications
2850°C | |
580°C | |
White | |
Solid | |
CaH2O2 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
[OH-].[OH-].[Ca++] | |
74.09 | |
CHEBI:31341 | |
Certified | |
Poly Bottle |
Calcium Hydroxide | |
1305-62-0 | |
12.4 | |
500 g | |
MFCD00010901 | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
calcium dihydroxide | |
6093208 | |
74.09 | |
Pass Test | |
2.240g/cm³ |
Chemical Identifiers
1305-62-0 | |
74.09 | |
AXCZMVOFGPJBDE-UHFFFAOYSA-L | |
6093208 | |
calcium dihydroxide |
CaH2O2 | |
MFCD00010901 | |
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid | |
CHEBI:31341 | |
[OH-].[OH-].[Ca++] |
Safety and Handling
WARNING!
Emergency Overview
Causes skin and eye burns. Causes respiratory tract irritation and possible burns. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:0
Instability:0
DOTInformation : DOT Class 8, : Corrosive