missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium ingot, 97% (metals basis), Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 01012839

Catalog No. AA1012839


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium
1.54
Ca
700 g
OYPRJOBELJOOCE-UHFFFAOYSA-N
calcium
5460341
40.078
7440-70-2
99% (metals basis)
MFCD00010897 MFCD00085314
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
[Ca]
40.08
CHEBI:29320
Ingot

Chemical Identifiers

7440-70-2
40.08
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
calcium
Ca
MFCD00010897 MFCD00085314
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
CHEBI:29320
[Ca]
Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H261

DOTInformation : Hazard Class: 4.3; Packaging Group: II

EINECSNumber : 213-179-5

RTECSNumber : EV8040000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only