missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium L-threonate, 98%, Thermo Scientific Chemicals

$148.10 - $432.81

Chemical Identifiers

CAS 70753-61-6
Molecular Formula C8H14CaO10
Molecular Weight (g/mol) 310.27
MDL Number MFCD00077882,MFCD00150824
InChI Key ZJXGOFZGZFVRHK-UHFFFAOYNA-L
Synonym calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
PubChem CID 13388558
IUPAC Name calcium;(2R,3S)-2,3,4-trihydroxybutanoate
SMILES [Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6627922
View Documents
Thermo Scientific Chemicals
J6627922
100 g
Each for $148.10
 
AAJ6627936
View Documents
Thermo Scientific Chemicals
J6627936
500 g
Each for $432.81
 
Description

Description

Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

Solubility
Soluble in water. Sparingly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

70753-61-6
310.27
ZJXGOFZGZFVRHK-UHFFFAOYNA-L
13388558
[Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
C8H14CaO10
MFCD00077882,MFCD00150824
calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
calcium;(2R,3S)-2,3,4-trihydroxybutanoate

Specifications

70753-61-6
Brown
MFCD00077882,MFCD00150824
calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
ZJXGOFZGZFVRHK-UHFFFAOYNA-L
calcium;(2R,3S)-2,3,4-trihydroxybutanoate
13388558
98%
Calcium L-threonate
>300°C
C8H14CaO10
100 g
Soluble in water. Sparingly soluble in methanol.
[Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
310.27
310.27
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only