missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium L-Threonate 98.0+%, TCI America™

Supplier:  TCI America T179925G

Catalog No. T179925G


Explore available promotions

Description

Description

Specifications

Specifications

Calcium L-Threonate
White
MFCD00077882,MFCD00150824
calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
[Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
310.27
310.27
Crystalline Powder
70753-61-6
C8H14CaO10
25 g
ZJXGOFZGZFVRHK-UHFFFAOYNA-L
calcium bis(2,3,4-trihydroxybutanoate)
13388558
≥98.0% (T)

Chemical Identifiers

70753-61-6
310.27
ZJXGOFZGZFVRHK-UHFFFAOYNA-L
13388558
[Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
C8H14CaO10
MFCD00077882,MFCD00150824
calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
calcium bis(2,3,4-trihydroxybutanoate)
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions