missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium metaborate, 99.992% (metals basis), Thermo Scientific Chemicals

$69.21 - $718.48

Chemical Identifiers

CAS 13701-64-9
Molecular Formula B2CaO4
Molecular Weight (g/mol) 125.694
MDL Number MFCD00049397
InChI Key ILOKQJWLMPPMQU-UHFFFAOYSA-N
Synonym calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
PubChem CID 10290800
IUPAC Name calcium;oxido(oxo)borane
SMILES B(=O)[O-].B(=O)[O-].[Ca+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1298806
View Documents
Thermo Scientific Chemicals
01298806
5 g
Each for $69.21
 
AA1298814
View Documents
Thermo Scientific Chemicals
01298814
25 g
Each for $224.19
 
AA1298822
View Documents
Thermo Scientific Chemicals
01298822
100 g
Each for $718.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13701-64-9
125.694
ILOKQJWLMPPMQU-UHFFFAOYSA-N
10290800
B(=O)[O-].B(=O)[O-].[Ca+2]
B2CaO4
MFCD00049397
calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
calcium;oxido(oxo)borane

Specifications

White
Powder
B2CaO4
MFCD00049397
ILOKQJWLMPPMQU-UHFFFAOYSA-N
calcium;oxido(oxo)borane
10290800
99.992%
Calcium metaborate
13701-64-9
5 g
Ca(BO2)2
calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
B(=O)[O-].B(=O)[O-].[Ca+2]
125.694
125.7
Odorless
(metals basis)
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-224-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only