Learn More
Calcium nitrate tetrahydrate, 99%, for analysis, Thermo Scientific Chemicals
$201.75 - $756.85
Chemical Identifiers
CAS | 13477-34-4 |
---|---|
Molecular Formula | CaN2O6·4H2O |
Molecular Weight (g/mol) | 236.15 |
MDL Number | MFCD00149604 |
InChI Key | ICSSIKVYVJQJND-UHFFFAOYSA-N |
Synonym | calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate |
PubChem CID | 16211656 |
ChEBI | CHEBI:86159 |
IUPAC Name | calcium;dinitrate;tetrahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC217510010
|
Thermo Scientific Chemicals
217510010 |
1 kg | Plastic bottle |
Each for $201.75
|
|
||||
AC217510050
|
Thermo Scientific Chemicals
217510050 |
5 kg | Plastic bottle |
Each for $756.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13477-34-4 | |
236.15 | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
16211656 | |
calcium;dinitrate;tetrahydrate |
CaN2O6·4H2O | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
CHEBI:86159 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Specifications
132.0°C | |
13477-34-4 | |
4.5 to 7.0 (5% soln. at 20°C) | |
1 kg | |
CaN2O6·4H2O | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
calcium;dinitrate;tetrahydrate | |
16211656 | |
236.15 | |
Analytical | |
1.86 | |
Calcium nitrate tetrahydrate |
45.0°C | |
White | |
Crystals | |
98.5% min. (Complexometry) | |
Ca(NO3)2·4H2O | |
15, 1684 | |
Solubility in water: 1470g/L (0°C). Other solubilities: soluble in alcohol and acetone | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] | |
236.15 | |
CHEBI:86159 | |
99% | |
Plastic bottle | |
1.8600g/mL |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May intensify fire; oxidizer.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO – Research Use Only