missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Nitrate Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical FLC109500
Specifications
132°C | |
45°C | |
White | |
Solid | |
99 to 103 % | |
Ca(NO3)2·4H2O | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] | |
236.146 | |
CHEBI:86159 | |
99 to 103% | |
Pass Test | |
1.860g/cm³ |
Calcium Nitrate Tetrahydrate | |
13477-34-4 | |
5 to 7 | |
500 g | |
CaH8N2O10 | |
MFCD00149604 | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
calcium;dinitrate;tetrahydrate | |
16211656 | |
236.15 | |
Certified ACS | |
Poly Bottle |
Chemical Identifiers
13477-34-4 | |
236.146 | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
16211656 | |
calcium;dinitrate;tetrahydrate |
CaH8N2O10 | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
CHEBI:86159 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Safety and Handling
DOTInformation : DOT Class 5.1, : Oxidizer