Learn More
Calcium oxide, Reagent Grade, Thermo Scientific Chemicals
$176.97 - $511.33
Chemical Identifiers
CAS | 1305-78-8 |
---|---|
Molecular Formula | CaO |
Molecular Weight (g/mol) | 56.08 |
MDL Number | MFCD00010911 |
InChI Key | BRPQOXSCLDDYGP-UHFFFAOYSA-N |
Synonym | calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx |
PubChem CID | 14778 |
ChEBI | CHEBI:31344 |
IUPAC Name | oxocalcium |
SMILES | [O--].[Ca++] |
Description
Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
1305-78-8 | |
56.08 | |
BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
14778 | |
oxocalcium |
CaO | |
MFCD00010911 | |
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
CHEBI:31344 | |
[O--].[Ca++] |
Specifications
2850°C | |
1305-78-8 | |
500 g | |
MFCD00010911 | |
Soluble in water with generation of heat,and forming Ca(OH)2,in acids,glycerol. Insoluble in alcoholSoluble in water. Insoluble in methanol,diethyl ether and n-octanol. | |
[O--].[Ca++] | |
56.08 | |
CHEBI:31344 | |
Reagent | |
Air and moisture sensitive | |
Calcium oxide |
2572°C | |
Powder | |
CaO | |
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx | |
BRPQOXSCLDDYGP-UHFFFAOYSA-N | |
oxocalcium | |
14778 | |
56.08 | |
1.83 | |
3.25-3.38 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 215-138-9
RTECSNumber : EW3100000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only