missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium phosphate, for analysis, 35-40% (Ca), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 213480010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium phosphate | |
c.p. | |
1 kg | |
Ca3O8P2 | |
15,1696 | |
Solubility in water: 0.1g/L (25°C) | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] | |
310.18 | |
CHEBI:9679 | |
35 to 40% (Ca) | |
Plastic bottle |
1670.0°C | |
7758-87-4 | |
35-40% | |
Ca3(PO4)2 | |
calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
tricalcium;diphosphate | |
24456 | |
310.18 | |
Analytical |
Chemical Identifiers
7758-87-4 | |
310.18 | |
calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic | |
CHEBI:9679 | |
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
Ca3O8P2 | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
24456 | |
tricalcium;diphosphate |
Safety and Handling
EINECSNumber : 231-840-8
TSCA : TSCA
RUO – Research Use Only