missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, dibasic dihydrate, 98+%, extra pure

CAS: 7789-77-7 | CaHO42H2O | 172.09 g/mol

$158.24 - $158.24

Chemical Identifiers

CAS 7789-77-7
Molecular Formula CaHO42H2O
Molecular Weight (g/mol) 172.09
MDL Number MFCD00149621
InChI Key XAAHAAMILDNBPS-UHFFFAOYSA-L
Synonym calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
PubChem CID 104805
ChEBI CHEBI:4496
IUPAC Name calcium;hydrogen phosphate;dihydrate
SMILES O.O.OP(=O)([O-])[O-].[Ca+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC389830010
SDS View Documents
Thermo Scientific Chemicals
389830010
1 kg Plastic bottle
Each for $158.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-77-7
172.09
XAAHAAMILDNBPS-UHFFFAOYSA-L
104805
calcium;hydrogen phosphate;dihydrate
CaHO42H2O
MFCD00149621
calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
CHEBI:4496
O.O.OP(=O)([O-])[O-].[Ca+2]

Specifications

White
Powder
98.0 to 105% (on dried substance) (Complexometry)
CaHPO4·2H2O
calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium;hydrogen phosphate;dihydrate
104805
172.09
Extra Pure
24.5% to 26.5% (800°C)
Calcium phosphate, dibasic dihydrate
7789-77-7
1 kg
CaHO42H2O
MFCD00149621
Solubility in water: 0.2g/L (25°C)
O.O.OP(=O)([O-])[O-].[Ca+2]
172.09
CHEBI:4496
≥98%
Calcium: positive, Phosphate: positive
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only