missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Phosphate Tribasic, MP Biomedicals

$78.97 - $577.30

Chemical Identifiers

CAS 12167-74-7
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.17
MDL Number MFCD00010904
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
PubChem CID 14781
IUPAC Name tricalcium diphosphate
SMILES [Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19380525
View Documents
MP Biomedicals Inc
0219380525
25 g
Each for $78.97
 
ICN19380580
View Documents
MP Biomedicals Inc
0219380580
100 g
Each for $222.47
 
ICN19380590
View Documents
MP Biomedicals Inc
0219380590
500 g
Each for $577.30
 
Description

Description

Specifications

Chemical Identifiers

12167-74-7
310.17
QORWJWZARLRLPR-UHFFFAOYSA-H
14781
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Ca3O8P2
MFCD00010904
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
tricalcium diphosphate

Specifications

12167-74-7
Ca3O8P2
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
310.17
502.3
25 g
MFCD00010904
QORWJWZARLRLPR-UHFFFAOYSA-H
tricalcium diphosphate
14781
Calcium Phosphate Tribasic
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'