missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Phosphate Tribasic, Powder, Spectrum™ Chemical
$497.29 - $11804.35
Chemical Identifiers
CAS | 1306-06-5 |
---|---|
Molecular Formula | Ca5HO13P3 |
Molecular Weight (g/mol) | 502.31 |
MDL Number | MFCD00010904 |
InChI Key | XYJRXVWERLGGKC-UHFFFAOYSA-D |
IUPAC Name | pentacalcium hydroxide triphosphate |
SMILES | [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18609523
|
Spectrum Chemical Mfg Cor
C1155125GM |
125 g | Poly Bottle |
Each for $497.29
|
|
||||
18609524
|
Spectrum Chemical Mfg Cor
C1155500GM |
500 g | Poly Bottle |
Each for $1,226.62
|
|
||||
18609525
|
Spectrum Chemical Mfg Cor
C115525KG |
2.5 kg | Poly Pail |
Each for $3,685.59
|
|
||||
18609526
|
Spectrum Chemical Mfg Cor
C115512KG |
12 kg | Poly Pail |
Each for $7,179.74
|
|
||||
18609527
|
Spectrum Chemical Mfg Cor
C115545KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1306-06-5 | |
502.31 | |
XYJRXVWERLGGKC-UHFFFAOYSA-D | |
[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
Ca5HO13P3 | |
MFCD00010904 | |
pentacalcium hydroxide triphosphate |
Specifications
1306-06-5 | |
125 g | |
MFCD00010904 | |
[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
502.31 | |
Poly Bottle |
1 | |
Ca5HO13P3 | |
XYJRXVWERLGGKC-UHFFFAOYSA-D | |
pentacalcium hydroxide triphosphate | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.