missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium pyruvate, 96%, Thermo Scientific Chemicals
$79.62 - $259.32
Chemical Identifiers
CAS | 52009-14-0 |
---|---|
Molecular Formula | C6H6CaO6 |
Molecular Weight (g/mol) | 214.186 |
MDL Number | MFCD00037199 |
InChI Key | UZWMCCLZMHPPKW-UHFFFAOYSA-L |
Synonym | calcium pyruvate, calcium 2-oxopropanoate, unii-9v2cqz19n4, 2-oxo-propanoic acid calcium salt, calcium pyruvate, anhydrous, propanoic acid, 2-oxo-, calcium salt, pyruvic acid calcium salt, calcium pyruvic, acmc-20ali9, calcium dipyruvate |
PubChem CID | 9859191 |
IUPAC Name | calcium;2-oxopropanoate |
SMILES | CC(=O)C(=O)[O-].CC(=O)C(=O)[O-].[Ca+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2520414
|
Thermo Scientific Chemicals
B2520414 |
25 g |
Each for $79.62
|
|
|||||
AAB2520422
|
Thermo Scientific Chemicals
B2520422 |
100 g |
Each for $259.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52009-14-0 | |
214.186 | |
UZWMCCLZMHPPKW-UHFFFAOYSA-L | |
9859191 | |
CC(=O)C(=O)[O-].CC(=O)C(=O)[O-].[Ca+2] |
C6H6CaO6 | |
MFCD00037199 | |
calcium pyruvate, calcium 2-oxopropanoate, unii-9v2cqz19n4, 2-oxo-propanoic acid calcium salt, calcium pyruvate, anhydrous, propanoic acid, 2-oxo-, calcium salt, pyruvic acid calcium salt, calcium pyruvic, acmc-20ali9, calcium dipyruvate | |
calcium;2-oxopropanoate |
Specifications
52009-14-0 | |
MFCD00037199 | |
calcium pyruvate, calcium 2-oxopropanoate, unii-9v2cqz19n4, 2-oxo-propanoic acid calcium salt, calcium pyruvate, anhydrous, propanoic acid, 2-oxo-, calcium salt, pyruvic acid calcium salt, calcium pyruvic, acmc-20ali9, calcium dipyruvate | |
CC(=O)C(=O)[O-].CC(=O)C(=O)[O-].[Ca+2] | |
214.186 | |
214.19 | |
Calcium pyruvate, anhydrous |
C6H6CaO6 | |
25 g | |
UZWMCCLZMHPPKW-UHFFFAOYSA-L | |
calcium;2-oxopropanoate | |
9859191 | |
97% |
Safety and Handling
EINECSNumber : 257-599-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only