Learn More
Calcium stearate, Thermo Scientific Chemicals
Waterproofing and release agent, stabilizer for PVC, lubricant, conditioning agent
Supplier: Thermo Scientific Chemicals 039423A1
Description
Used as a stabilizer and a lubricant in rigid PVC processing, PVC compounding and moulding, in leather cloth manufacturing. It is used as a lubricant, in iron wire drawing and used as an anti caking agent in detergent Cakes and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium stearate | |
179°C to 180°C | |
MFCD00036390 | |
14,1703 | |
Soluble in hot pyridine,slightly soluble in oil,but insoluble in alcohol and ether. | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2] | |
607.03 | |
607.04 |
1592-23-0 | |
C36H70CaO4 | |
1 kg | |
calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates | |
CJZGTCYPCWQAJB-UHFFFAOYSA-L | |
calcium;octadecanoate | |
15324 | |
Powder |
Chemical Identifiers
1592-23-0 | |
607.03 | |
CJZGTCYPCWQAJB-UHFFFAOYSA-L | |
15324 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2] |
C36H70CaO4 | |
MFCD00036390 | |
calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates | |
calcium;octadecanoate |
Safety and Handling
EINECSNumber : 216-472-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only