missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium sulfate, 99%, pure, anhydrous, Thermo Scientific Chemicals
$100.89 - $1104.66
Chemical Identifiers
CAS | 7778-18-9 |
---|---|
Molecular Formula | CaO4S |
Molecular Weight (g/mol) | 136.13 |
MDL Number | MFCD00010912 |
InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
Synonym | calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous |
PubChem CID | 24497 |
ChEBI | CHEBI:31346 |
IUPAC Name | calcium;sulfate |
SMILES | [Ca++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC217521000
|
Thermo Scientific Chemicals
217521000 |
100 g | Plastic bottle |
Each for $100.89
|
|
|||||
AC217525000
|
Thermo Scientific Chemicals
217525000 |
500 g | Plastic bottle |
Each for $272.92
|
|
|||||
AC217520025
|
Thermo Scientific Chemicals
217520025 |
2.5 kg | Plastic bottle |
Each for $1,104.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentChemical Identifiers
7778-18-9 | |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
24497 | |
calcium;sulfate |
CaO4S | |
MFCD00010912 | |
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous | |
CHEBI:31346 | |
[Ca++].[O-]S([O-])(=O)=O |
Specifications
1450.0°C | |
White | |
100 g | |
CaO4S | |
MFCD00010912 | |
15,1708 | |
Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
CHEBI:31346 | |
99% | |
Plastic bottle |
7778-18-9 | |
Fine Crystalline Powder | |
98.5% min | |
CaSO4 | |
01,107 | |
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium;sulfate | |
24497 | |
136.14 | |
Pure | |
Calcium sulfate |
Safety and Handling
EINECSNumber : 231-900-3
RUO – Research Use Only