missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium sulfate dihydrate, 98+%, ACS reagent
CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol
$256.32 - $831.90
Chemical Identifiers
| CAS | 10101-41-4 |
|---|---|
| Molecular Formula | CaH4O6S |
| Molecular Weight (g/mol) | 172.16 |
| MDL Number | MFCD00149625 |
| InChI Key | PASHVRUKOFIRIK-UHFFFAOYSA-L |
| Synonym | calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone |
| PubChem CID | 24928 |
| ChEBI | CHEBI:32583 |
| IUPAC Name | calcium;sulfate;dihydrate |
| SMILES | O.O.[Ca++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC315255000
|
Thermo Scientific Chemicals
315255000 |
500 g | Plastic bottle |
N/A
|
|
||||
|
AC315250025
|
Thermo Scientific Chemicals
315250025 |
2.5 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10101-41-4 | |
| 172.16 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| 24928 | |
| calcium;sulfate;dihydrate |
| CaH4O6S | |
| MFCD00149625 | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| CHEBI:32583 | |
| O.O.[Ca++].[O-]S([O-])(=O)=O |
Specifications
| 10101-41-4 | |
| Powder | |
| 98.0 to 102% (Complexometry) | |
| CaSO4·2H2O | |
| 01,107 | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| calcium;sulfate;dihydrate | |
| 24928 | |
| 172.17 | |
| ACS Reagent | |
| Calcium sulfate dihydrate |
| White | |
| 500 g | |
| CaH4O6S | |
| MFCD00149625 | |
| 14, 1706 | |
| Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents | |
| O.O.[Ca++].[O-]S([O-])(=O)=O | |
| 172.16 | |
| CHEBI:32583 | |
| ≥98% | |
| Plastic bottle |
Safety and Handling
RTECSNumber : EW4150000
RUO – Research Use Only