missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium thiosulfate, pure, 30-50% solution in water, Thermo Scientific Chemicals

$614.13 - $614.13

Specifications

Chemical Name or Material Calcium thiosulfate
CAS 10124-41-1,7732-18-5
Molecular Formula CaO3S2
Linear Formula CaS2O3
Synonym calcium thiosulfate, tecesal, calcium thiosulphate, calcium hyposulfite, caswell no. 153, unii-2io10o728e, calcium thiosulfate ca s2o3, epa pesticide chemical code 080101, thiosulfuric acid h2s2o3 , calcium salt 1:1, thiosulfuric acid, calcium salt 1:1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC447870050
View Documents
Thermo Scientific Chemicals
447870050
5 L
Each for $614.13
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium thiosulfate
CaO3S2
calcium thiosulfate, tecesal, calcium thiosulphate, calcium hyposulfite, caswell no. 153, unii-2io10o728e, calcium thiosulfate ca s2o3, epa pesticide chemical code 080101, thiosulfuric acid h2s2o3 , calcium salt 1:1, thiosulfuric acid, calcium salt 1:1
FAYYUXPSKDFLEC-UHFFFAOYSA-L
calcium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
24964
Pure
1.2450g/mL
10124-41-1,7732-18-5
CaS2O3
Solubility in water: soluble
[O-]S(=O)(=S)[O-].[Ca+2]
152.21
152.21
1.245
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only