missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium zirconium oxide, 99.2% (metals basis), Thermo Scientific Chemicals
$75.29 - $196.68
Chemical Identifiers
CAS | 12013-47-7 |
---|---|
Molecular Formula | CaO3Zr |
Molecular Weight (g/mol) | 179.299 |
MDL Number | MFCD00015982 |
InChI Key | BJJVDFHQHSBAFC-UHFFFAOYSA-N |
Synonym | calcium zirconate, calcium oxozirconiumbis olate, calcium;dioxido oxo zirconium, calcium 2+ oxozirconiumbis olate |
PubChem CID | 16212531 |
IUPAC Name | calcium;dioxido(oxo)zirconium |
SMILES | [O-][Zr](=O)[O-].[Ca+2] |
Description
Calcium zirconium oxide is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12013-47-7 | |
179.299 | |
BJJVDFHQHSBAFC-UHFFFAOYSA-N | |
16212531 | |
[O-][Zr](=O)[O-].[Ca+2] |
CaO3Zr | |
MFCD00015982 | |
calcium zirconate, calcium oxozirconiumbis olate, calcium;dioxido oxo zirconium, calcium 2+ oxozirconiumbis olate | |
calcium;dioxido(oxo)zirconium |
Specifications
2550°C | |
Powder | |
CaO3Zr | |
calcium zirconate, calcium oxozirconiumbis olate, calcium;dioxido oxo zirconium, calcium 2+ oxozirconiumbis olate | |
BJJVDFHQHSBAFC-UHFFFAOYSA-N | |
calcium;dioxido(oxo)zirconium | |
16212531 | |
99.2% | |
Calcium zirconium oxide |
12013-47-7 | |
250 g | |
MFCD00015982 | |
Soluble in Nitric acid. | |
[O-][Zr](=O)[O-].[Ca+2] | |
179.299 | |
179.3 | |
Odorless | |
(metals basis) |
Safety and Handling
EINECSNumber : 234-586-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only