missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Calconcarboxylic acid, indicator grade

Calconcarboxylic acid, indicator grade, Acros Organics, Quantity: 250g, Packaging: Glass bottle, Black to Purple, Grade: Extra Pure, Melting Point: 300 deg.C, Molecular Weight: 438.4, CAS: 3737-95-9, EINECS Number: 223-117-, Formula Weight: 438.4, Health Hazard 1: GHS Signal Word: Warning

$100.66 - $306.16

Chemical Identifiers

CAS 3737-95-9
Molecular Formula C21H14N2O7S
Molecular Weight (g/mol) 438.4
MDL Number MFCD00004078
InChI Key ULIVOAKVRBXKKS-PYCFMQQDSA-N
Synonym calconcarboxylic acid, patton-reeder indicator, calconcarbonic acid, calcon 3-carboxylic acid, patton and reeder's indicator, 2-hydroxy-1-2-hydroxy-4-sulfo-1-napthylazo-3-naphthoic acid, 2,2'-dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy-4-2-hydroxy-4-sulfo-1-naphthalenyl azo, nn
PubChem CID 5895210
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC157310100
SDS View Documents
Thermo Scientific Chemicals
157310100
10 g Glass bottle
Each for $100.66
 
AC157310500
SDS View Documents
Thermo Scientific Chemicals
157310500
50 g Glass bottle
Each for $306.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

3737-95-9
438.4
ULIVOAKVRBXKKS-PYCFMQQDSA-N
5895210
C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O
C21H14N2O7S
MFCD00004078
calconcarboxylic acid, patton-reeder indicator, calconcarbonic acid, calcon 3-carboxylic acid, patton and reeder's indicator, 2-hydroxy-1-2-hydroxy-4-sulfo-1-napthylazo-3-naphthoic acid, 2,2'-dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy-4-2-hydroxy-4-sulfo-1-naphthalenyl azo, nn
3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid

Specifications

3737-95-9
Black to Purple
Glass bottle
MFCD00004078
calconcarboxylic acid, patton-reeder indicator, calconcarbonic acid, calcon 3-carboxylic acid, patton and reeder's indicator, 2-hydroxy-1-2-hydroxy-4-sulfo-1-napthylazo-3-naphthoic acid, 2,2'-dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy-4-2-hydroxy-4-sulfo-1-naphthalenyl azo, nn
ULIVOAKVRBXKKS-PYCFMQQDSA-N
3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
5895210
Extra Pure
Calconcarboxylic acid
300°C
Authentic
C21H14N2O7S
10 g
Solubility in water: slightly soluble. Other solubilities: soluble in methanol and ethanol
C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O
438.4
438.4
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 223-117-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only