BINDER™ Calibration Certificate 1 P Temperature and Humidity for Series KBF-S, KBF-S ECO
BINDER™ Calibration certificate is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
Supplier: BINDER™ 80122065
Description
- Calibration certificate, temperature and humidity
- Measurement in center of chamber at 25°C / 60% RH or at specified test values
Specifications
BINDER Chambers Series KBF-S ECO | |
Calibration certificate, temperature and humidity - Measurement in center of chamber at 25°C / 60% RH or at specified test values | |
1 Day | |
Calibration Certificate |