missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Calibrex™ organo 525 Bottle Top Dispensers

Adjustable dispensers include a ground glass plunger that is best suited for organic, non-crystallizing acid and base solutions.

Supplier:  DWK Life Sciences W845002

 View more versions of this product

Catalog No. 03-410-901


Explore available promotions

Description

Description

  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Specifications

Specifications

Bottletop Dispenser
0.25 to 2.5 mL
Calibrex organo 525
Autoclavable
Analog
2 Years
Videos
SDS
Documents

Documents

Promotions

Promotions