DWK Life Sciences Calibrex™ solutae 530 Bottle Top Dispenser
Fits most laboratory bottles either directly or using supplied adapters. Calibrex™ solutae 530 Bottle Top Dispensers feature excellent chemical resistance, high volumetry performance and simplified maintenance. Volume range stretches between 0.1 and
$1950.12 - $1950.12
Specifications
Product Type Bottletop Dispenser
Autoclavable Autoclavable
For Use With (Equipment) Fits most laboratory bottles directly or with supplied adapters
Includes PFA-coated plunger and integrated calibration
Model Calibrex solutae 530
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
03-410-910
View Documents Promotion Details
DWK Life Sciences
W845020
0.5 to 5 mL
Case for $1,950.12
Description
Description
The Calibrex™ 530 solutae has a PFA-coated plunger preventing the plunger and barrel from seizing together. This enables trouble free distribution of salt solutions, weak and strong acids, as well as bases. The materials in contact with liquid flow are chemically inert, providing for stability and long instrument life.
  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Specifications
Specifications
Bottletop Dispenser
Fits most laboratory bottles directly or with supplied adapters
Calibrex solutae 530
Autoclavable
PFA-coated plunger and integrated calibration
2 Years
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'