Learn More
Calix[4]arene, 98%, Thermo Scientific Chemicals
$206.36 - $637.34
Chemical Identifiers
CAS | 74568-07-3 |
---|---|
Molecular Formula | C28H24O4 |
Molecular Weight (g/mol) | 424.496 |
MDL Number | MFCD00143912 |
InChI Key | YPNHVQZZPXPQOS-UHFFFAOYSA-N |
Synonym | calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc |
PubChem CID | 562409 |
SMILES | C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2132003
|
Thermo Scientific Chemicals
B2132003 |
1 g |
N/A
|
|
||||||
AAB2132006
|
Thermo Scientific Chemicals
B2132006 |
5 g |
N/A
|
|
||||||
Description
Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFunctionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,
Solubility
Soluble in chloroform.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
74568-07-3 | |
424.496 | |
YPNHVQZZPXPQOS-UHFFFAOYSA-N | |
562409 |
C28H24O4 | |
MFCD00143912 | |
calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc | |
C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O |
Specifications
74568-07-3 | |
C28H24O4 | |
1 g | |
Soluble in chloroform. | |
C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O | |
562409 | |
98% |
315°C to 318°C | |
MFCD00143912 | |
calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc | |
YPNHVQZZPXPQOS-UHFFFAOYSA-N | |
424.496 | |
424.5 | |
Calix[4]arene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only