missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Calmagite, indicator grade

Supplier:  Thermo Scientific Chemicals B2148206

 View more versions of this product

Catalog No. AAB2148206

Explore available promotions

Description

Description

Calmagite is widely used in analytical chemistry as an indicator in the titration of calcium or magnesium with ethylenediaminetetraacetic acid (EDTA). It is also useful in the determination of magnesium in biological materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calmagite is widely used in analytical chemistry as an indicator in the titration of calcium or magnesium with ethylenediaminetetraacetic acid (EDTA). It is also useful in the determination of magnesium in biological materials.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Calmagite
3147-14-6
MFCD00011656
14,1718
ASFVMSDYPYMUFL-UHFFFAOYSA-N
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
6364506
Indicator
>330°C
C17H14N2O5S
708596
Soluble in water.
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
358.368
358.37
5 g

Chemical Identifiers

3147-14-6
358.368
ASFVMSDYPYMUFL-UHFFFAOYSA-N
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
C17H14N2O5S
MFCD00011656
6364506
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 221-563-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only