Cambridge Diagnostic Products, Inc.™ Camco Quik Stain™ and Quik Stain™ II Edge
Edge
Single, easy-to-use solutions provide fast and consistent differential staining of all cell types in less than 30 seconds
Supplier:  Cambridge Diagnostic Products, Inc.™ 211
 View more versions of this product
Catalog No. 04-330-12
Edge
Explore available promotions
Description
Description
  • Both are self-buffering and intensity controlled by washing with ordinary distilled water
  • CAMCO Quik Stain is a Wright's stain formulation (traditional blue coloration) and CAMCO Quik Stain II is a Wright-Giemsa stain (traditional coloration with blue-red highlights)
Specifications
Specifications
946 mL
Liquid
Dark Blue
64°C
Quik Stain II