Cambridge Diagnostic Products, Inc.™ Sequester-Sol™ Anticoagulant
For in vitro diagnostic use
Supplier:  Cambridge Diagnostic Products, Inc.™ 205
Catalog No. 23564252
Explore available promotions
Description
Description
  • Liquid form, non toxic anticoagulant
  • For hematology, blood chemistry procedures
  • Gives complete blood counts, blood smears for staining within 48 hours
  • One drop per 5mL of blood
  • Comes with dropper tip for accurate dispensing
  • Packed in polyethylene bottle
  • Sufficient for hundreds of samples
  • Convenient and economical to use
For hematology, blood chemistry procedures.
Specifications
Specifications
Anticoagulant