Promega cAMP-Glo™ Assay DFS Item
The cAMP-Glo™ Assay is a homogeneous, bioluminescent and high-throughput assay for measuring cAMP levels in cells
Supplier:  Promega V1501
 View more versions of this product
Catalog No. PRV1501
Explore available promotions
Description
Description
  • Simple, Bioluminescent cAMP Detection
  • Complete in ∼45 minutes
  • Scalable to 1536-well plate formats and beyond
  • No interference by fluorescent compounds
Specifications
Specifications
300 Assays
384-well Plate
For 384-well plates
-30°C to -10°C
For high throughput analysis of library compounds to identify modulators of GPCR activity