Learn More
Camphene, 75%, remainder mainly α-fenchene, Thermo Scientific Chemicals
$574.88 - $574.88
Chemical Identifiers
CAS | 79-92-5 |
---|---|
Molecular Formula | C10H16 |
Molecular Weight (g/mol) | 136.24 |
MDL Number | MFCD00066603 |
InChI Key | CRPUJAZIXJMDBK-UHFFFAOYSA-N |
Synonym | camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 |
PubChem CID | 6616 |
ChEBI | CHEBI:3830 |
IUPAC Name | 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane |
SMILES | CC1(C2CCC(C2)C1=C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC295570025
|
Thermo Scientific Chemicals
295570025 |
2.5 L | Glass bottle |
Each for $574.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-92-5 | |
136.24 | |
CRPUJAZIXJMDBK-UHFFFAOYSA-N | |
6616 | |
3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane |
C10H16 | |
MFCD00066603 | |
camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 | |
CHEBI:3830 | |
CC1(C2CCC(C2)C1=C)C |
Specifications
79-92-5 | |
White | |
36°C | |
70% min. (GC) | |
C10H16 | |
2.5 L | |
15, 1732 | |
Solubility in water: negligible. Other solubilities: soluble in ether,cyclohexane,cyclohexene,,dioxane and chloroform,slightly soluble in alcohol | |
CC1(C2CCC(C2)C1=C)C | |
136.24 | |
1.6 mPa.s (50°C) | |
136.24 | |
Crystalline Low Melting Solid |
42°C | |
159.0°C to 160.0°C | |
Authentic | |
Glass bottle | |
MFCD00066603 | |
05, 156 | |
camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 | |
CRPUJAZIXJMDBK-UHFFFAOYSA-N | |
3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane | |
6616 | |
CHEBI:3830 | |
75% | |
Camphene, 75% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Flammable solid.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
If eye irritation persists: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiti
GHS Signal Word: Warning
EINECSNumber : 201-234-8
RTECSNumber : EX1055000
TSCA : TSCA
RUO – Research Use Only