missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(+/-)-Camphorquinone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1496714
Description
Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCamphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.
Solubility
Soluble in ethanol, ethyl ether and benzene.
Notes
Incompatible with strong oxidizing agents.
Specifications
(+/-)-Camphorquinone | |
198°C to 202°C | |
MFCD00064160 | |
1909463 | |
Soluble in ethanol,ethyl ether and benzene. | |
CC1(C2CCC1(C(=O)C2=O)C)C | |
166.22 | |
CHEBI:34607 | |
99% |
10373-78-1 | |
C10H14O2 | |
25 g | |
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone | |
VNQXSTWCDUXYEZ-UHFFFAOYSA-N | |
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione | |
25208 | |
166.22 |
Chemical Identifiers
10373-78-1 | |
166.22 | |
VNQXSTWCDUXYEZ-UHFFFAOYSA-N | |
25208 | |
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione |
C10H14O2 | |
MFCD00064160 | |
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone | |
CHEBI:34607 | |
CC1(C2CCC1(C(=O)C2=O)C)C |
Safety and Handling
EINECSNumber : 233-814-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only