missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-Camphorquinone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1496722

 View more versions of this product

Catalog No. AAA1496722


Explore available promotions

Description

Description

Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

Solubility
Soluble in ethanol, ethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

(+/-)-Camphorquinone
198°C to 202°C
MFCD00064160
1909463
Soluble in ethanol,ethyl ether and benzene.
CC1(C2CCC1(C(=O)C2=O)C)C
166.22
CHEBI:34607
99%
10373-78-1
C10H14O2
100 g
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
25208
166.22

Chemical Identifiers

10373-78-1
166.22
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
25208
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
C10H14O2
MFCD00064160
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
CHEBI:34607
CC1(C2CCC1(C(=O)C2=O)C)C
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-814-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only