missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Canada Balsam

For microscopy. Commonly used mounting medium to prepare permanent slides for microscopy. It is produced from the resin of the balsam fir tree and its use can be combined with xylene-containing specimens.

Supplier:  MilliporeSigma™ 1016910100

 View more versions of this product

Catalog No. M1016910100


Explore available promotions

Description

Description

  • Density: 0.98g/cm3 (20°C)
  • CAS Registry Number: 8007-47-4
Specifications

Specifications

Glass Bottle
5° to 30°C
100 mL
Videos
SDS
Documents

Documents

Promotions

Promotions