missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cancer Diagnostics, Inc.™ Moist Mark Plus™ Marking Pen DFS Item

$150.32 - $1527.76

Specifications

Product Type Marking Pen
For Use With (Application) Useful in cytology as a “dotting” reference
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
22-500-210 DFS Item
View Documents
Cancer Diagnostics, Inc.™
MP1000DUP
10 Pk.
Pack of 10 Each for $150.32
 
22-500-465 DFS Item
View Documents
Cancer Diagnostics, Inc.™
MP2100
100 Cs.
Case of 100 for $1,527.76
 
Description

Description

  • Premium slide/cassette marker
  • Writes smooth, dark, and fine on all cassettes and slides
  • Useful in cytology as a “dotting” reference

Specifications

Specifications

Marking Pen
Useful in cytology as a “dotting” reference
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'