Cadence Science™ Cancer Implant Needle
Needle provides simple procedure to obtain and implant tumorous tissue cells beneath skin of lab animals for observation while maintaining a sterile field
$3595.91 - $3595.91
Specifications
For Use With (Application) Obtain and implant tumorous tissue cells beneath the skin of lab animals
Type Cancer Implant Needle
Material Stainless Steel
Size Standard
Length (English) 3.25 in.
View More Specs
Products 1
Catalog Number Mfr. No. Thickness Price Quantity  
Catalog Number Mfr. No. Thickness Price Quantity  
14-825-284
View Documents
Cadence Science™
POP7927
13 ga
Dozen of 12 Each for $3,595.91
Description
Description
  • Two piece needle set
  • Consists of sharp pointed stainless-steel outer cannula and stainless steel blunt obturator which protrudes 3.2mm beyond the needle point
Specifications
Specifications
Obtain and implant tumorous tissue cells beneath the skin of lab animals
Stainless Steel
3.25 in.
Cancer Implant Needle
Standard
8.25 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'