Cannon™ Fenske Opaque Viscometer
CFO Calibrated Series that determines viscosities using ASTM testing methods.
$757.14 - $757.14
Specifications
Product Type Cannon-Fenske Opaque Viscometer
Calibrated Yes
Certifications/Compliance ASTM D445, ASTM D446, ISO 3104, ISO 3105, calibration certificate
Includes Instruction sheet, calibration certificate
Material Borosilicate Glass
View More Specs
Includes: Certificate of calibration and instruction sheet
Products 1
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Range Price Quantity  
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Range Price Quantity  
22-273-946
View Documents
N/A 8 cSt/sec. 500 500 1600 to 8000 cSt
Each for $757.14
Description
Description
Permit measurements of liquids whose meniscus cannot be seen at the timing marks when using Cannon-Fenske Routine-type viscometer
  • Used to measure kinematic viscosity of liquid (cutback) asphalts and road oils at 60°C (140°F) in range of 30 to 6000 centistokes according to ASTM D 2170
  • Also used to study lubricating oils at low temperatures when investigating the effect of various additives on lubricating and hydraulic oils
  • External pressure can be applied to vary shear stress and shear rate
  • Upper bulb may be used for determining the degree of non-Newtonian behavior of the sample
Compliance
Specifications conform to ASTM D 446 and ISO 3105 for single determinations of viscosity for opaque liquids
Specifications
Specifications
Cannon-Fenske Opaque Viscometer
ASTM D445, ASTM D446, ISO 3104, ISO 3105, calibration certificate
Borosilicate Glass
22.9 cm
For measurement of kinematic viscosity of dark Newtonian liquids, measure kinematic viscosity of liquid and to study lubricating oils, upper bulb may be used for determining the degree of non-Newtonian behavior of the sample
Yes
Instruction sheet, calibration certificate
9 in.
±0.3%
12 mL
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'