missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Canrenone, 98%, Thermo Scientific Chemicals

Inhibits aldosterone biosynthesis and blocks ouabain effects

Supplier:  Thermo Scientific Chemicals J6023806

 View more versions of this product

Catalog No. AAJ6023806


Explore available promotions

Description

Description

Inhibits aldosterone biosynthesis and blocks ouabain effects

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Canrenone
149°C to 151°C
MFCD00071735
14,1752
UJVLDDZCTMKXJK-WNHSNXHDSA-N
(8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione
13789
98%
976-71-6
C22H28O3
5 g
canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone
CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C
340.463
340.46
Solid

Chemical Identifiers

976-71-6
340.463
UJVLDDZCTMKXJK-WNHSNXHDSA-N
13789
CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C
C22H28O3
MFCD00071735
canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone
(8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-554-5

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only