missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Canrenone, 98%, Thermo Scientific Chemicals
Inhibits aldosterone biosynthesis and blocks ouabain effects
Supplier: Thermo Scientific Chemicals J6023814
Description
Inhibits aldosterone biosynthesis and blocks ouabain effects
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Canrenone | |
149°C to 151°C | |
MFCD00071735 | |
14,1752 | |
UJVLDDZCTMKXJK-WNHSNXHDSA-N | |
(8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione | |
13789 | |
98% |
976-71-6 | |
C22H28O3 | |
25 g | |
canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C | |
340.463 | |
340.46 | |
Solid |
Chemical Identifiers
976-71-6 | |
340.463 | |
UJVLDDZCTMKXJK-WNHSNXHDSA-N | |
13789 | |
CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C |
C22H28O3 | |
MFCD00071735 | |
canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone | |
(8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione |
Safety and Handling
EINECSNumber : 213-554-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only