MilliporeSigma™ Supelco™ Capillary Starter Kit
Includes all of the necessary tools for installing capillary GC columns and related accessories
Supplier:  MilliporeSigma™ Supelco™ 23639
Catalog No. 111009397
Explore available promotions
Description
Description
Specifications
Specifications
Installing Capillary GC Columns and Related Accessories
Pipe Cleaners (6 in.), Tweezers, Pocket Mirror with Rotating Head, Open End Wrench (1/4 x 5/16 in.), Capillary Cleaving Tool, Stainless Steel Ruler (6 in.), Small Flashlight, Pin Vise Drill Kit, and Screw-Type Septum Puller
1 Kit
Capillary Starter Kit