3M™ DBI-Sala™ Delta™ Vest Style Harnesses
No-Tangle™ triangular design creates frame that allows the harness to fall into place when donned.
Supplier:  3M™ 1102008C
 View more versions of this product
Catalog No. 19-809-339
Explore available promotions
Description
Description
  • Spring-loaded stand-up dorsal D-ring design ensures fast, easy and safe connections
  • Rip-stitch indicators allow to inspect the harness at glance, for prior damaging impact loads
  • Harness falls into place, allowing quick and easy donning
  • Side D-rings for positioning
  • Adjustable non-slip chest strap with easy to use pass-thru buckle
  • Tongue buckle leg straps for ease of use and will not loose
  • Lanyard keepers holds unused snap hooks and help reduce trip hazard
  • Revolver vertical torso adjuster provides simple and fast adjustment eliminating loose ends and locks webbing into place preventing slippage
  • Velcro cover protects harness labels and holds I-Safe™ ID tag
Compliance
Meets ANSI™ Z359.11 requirements.
Specifications
Specifications
Tongue Buckle
Pass-Through
Polyester Webbing
Universal
Vest Style
1 back, 2 hip