3M™ DBI-Sala™ ExoFit NEX™ Vest-Style Positioning/Climbing Harness
For comfort, security and no-tangle donning.
Supplier:  3M™ 7012755758
 View more versions of this product
Catalog No. 19-810-559
Explore available promotions
Description
Description
  • Hybrid shoulder, back and leg comfort padding
  • Stand-up Tech-Lite aluminum back D-ring
  • Tech-Lite aluminum front D-ring
  • Tech-Lite aluminum side D-rings
  • Duo-Lok quick connect chest and leg straps
  • Repel webbing
  • Protective shoulder caps
  • Loops for body belt
  • Built-in lanyard keepers
  • Revolver vertical torso adjusters
  • Impact Indicator
  • Integrated trauma straps
  • Built-in reflective materials
  • Protected labels, equipped with i-Safe
Compliance
Meets OSHA 1910.66 and OSHA 1926.502; ANSI™ A10.32, ANSI Z359.1, ANSI Z359.3 and ANSI Z359.4 requirements.
Specifications
Specifications
Large