3M™ DBI-Sala™ Salalift II Winch and Tripod Rescue Systems
With overload clutch that reduces chance of injury to entangled worker
Supplier:  3M™ 8300030
 View more versions of this product
Catalog No. 19-809-578
Explore available promotions
Description
Description
  • Durable and lightweight corrosion resistant construction
  • With anodized aluminum side plates, zinc plated gears and shafts, stainless steel springs and aluminum drum
  • UL classified 7 in. (17.8cm) aluminum tripod
  • Quick mount bracket for winch attachment
  • Working load rating: 350 lb. (158.8kg) for work support and 310 lb. (140.6kg) for fall arrest
  • For raising, lowering, work support or personnel rescue
  • Comes with winch bag
  • With choice of 60 ft. (18.3m) of 0.25 in. (0.6cm) galvanized or stainless-steel cables
Specifications
Specifications
7 ft. Aluminum tripod, winch salafit II, quick mount bracket, 60 ft. L of 0.25 in. dia. Galvanized Steel cable
Ideal for raising, lowering, work support or personnel rescue
UL