missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ε-Caprolactam, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 108230025

 View more versions of this product

Catalog No. AC108230025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

ε-Caprolactam
68°C to 71°C
268°C
Authentic
Plastic bottle
MFCD00006936
21, V,6, 444
15, 1763
Solubility in water: 4560g/L (20°C). Other solubilities: freely soluble in methanol,dimethylformamide,,ethanol,ether and tetrahydrofurfuryl alcohol,soluble in chlorinated hydrocarbons,cyclohexene,petroleum fractions,chloroform,benzene
C1CCC(=O)NCC1
113.16
8.52 mPa.s (80°C)
113.16
Crystals or Flakes
105-60-2
White
152°C
99% min. (GC)
C6H11NO
2.5 kg
09,316
epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam
JBKVHLHDHHXQEQ-UHFFFAOYSA-N
azepan-2-one
7768
CHEBI:28579
99+%

Chemical Identifiers

105-60-2
113.16
JBKVHLHDHHXQEQ-UHFFFAOYSA-N
7768
azepan-2-one
C6H11NO
MFCD00006936
epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam
CHEBI:28579
C1CCC(=O)NCC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-313-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only