Learn More
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 108232500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
ε-Caprolactam | |
68°C to 71°C | |
268°C | |
Authentic | |
Plastic bottle | |
MFCD00006936 | |
21, V,6, 444 | |
15, 1763 | |
Solubility in water: 4560g/L (20°C). Other solubilities: freely soluble in methanol,dimethylformamide,,ethanol,ether and tetrahydrofurfuryl alcohol,soluble in chlorinated hydrocarbons,cyclohexene,petroleum fractions,chloroform,benzene | |
C1CCC(=O)NCC1 | |
113.16 | |
8.52 mPa.s (80°C) | |
113.16 | |
Crystals or Flakes |
105-60-2 | |
White | |
152°C | |
99% min. (GC) | |
C6H11NO | |
250 g | |
09,316 | |
epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam | |
JBKVHLHDHHXQEQ-UHFFFAOYSA-N | |
azepan-2-one | |
7768 | |
CHEBI:28579 | |
99+% |
Chemical Identifiers
105-60-2 | |
113.16 | |
JBKVHLHDHHXQEQ-UHFFFAOYSA-N | |
7768 | |
azepan-2-one |
C6H11NO | |
MFCD00006936 | |
epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam | |
CHEBI:28579 | |
C1CCC(=O)NCC1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-313-2
RUO – Research Use Only