missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPS, 0.5M buffer soln., pH 9.0, Thermo Scientific™
$102.22 - $102.22
Specifications
| CAS | 1135-40-6 |
|---|---|
| Concentration | 0.5 M |
| pH | 9.0 |
| Physical Form | Liquid |
| Molecular Formula | C9H19NO3S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63749AK
|
Thermo Scientific Chemicals
J63749AK |
250 mL |
Each for $102.22
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1135-40-6 | |
| 9.0 | |
| C9H19NO3S | |
| PJWWRFATQTVXHA-UHFFFAOYSA-N | |
| 3-(cyclohexylamino)propane-1-sulfonic acid |
| 0.5 M | |
| Liquid | |
| MFCD00003837 | |
| OS(=O)(=O)CCCNC1CCCCC1 | |
| 221.32 |