Learn More
Capsazepine, 98+%, Thermo Scientific Chemicals
A competitive antagonist for capsaicin and resiniferatoxin
$556.99 - $2416.03
Chemical Identifiers
CAS | 138977-28-3 |
---|---|
Molecular Formula | C19H21ClN2O2S |
Molecular Weight (g/mol) | 376.899 |
MDL Number | MFCD00153778 |
InChI Key | DRCMAZOSEIMCHM-UHFFFAOYSA-N |
Synonym | capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464 |
PubChem CID | 2733484 |
ChEBI | CHEBI:70773 |
IUPAC Name | N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide |
SMILES | C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ63055MA
|
Thermo Scientific Chemicals
J63055MA |
10 mg |
Each for $556.99
|
|
|||||
AAJ63055MF
|
Thermo Scientific Chemicals
J63055MF |
50 mg |
Each for $2,416.03
|
|
|||||
Description
Capsazepine is shown to inhibit the development of mechanical hyperalgesia induced by intraplanar capsaicin injection. Capsazepine is described to block Resiniferatoxin (sc-24015)- and Capsaicin-induced contractions of guinea pig treacheal smooth muscle. Studies investigating the function of the VR1 (TRPV1 receptor) have employed Capsazepine as a tool for perturbing systemic neuronal response to Capsaicin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCapsazepine is shown to inhibit the development of mechanical hyperalgesia induced by intraplanar capsaicin injection. Capsazepine is described to block Resiniferatoxin (sc-24015)- and Capsaicin-induced contractions of guinea pig treacheal smooth muscle. Studies investigating the function of the VR1 (TRPV1 receptor) have employed Capsazepine as a tool for perturbing systemic neuronal response to Capsaicin.
Solubility
Soluble in ethanol or DMSO
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Chemical Identifiers
138977-28-3 | |
376.899 | |
DRCMAZOSEIMCHM-UHFFFAOYSA-N | |
2733484 | |
N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide |
C19H21ClN2O2S | |
MFCD00153778 | |
capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464 | |
CHEBI:70773 | |
C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O |
Specifications
138977-28-3 | |
White | |
MFCD00153778 | |
capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464 | |
DRCMAZOSEIMCHM-UHFFFAOYSA-N | |
N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide | |
2733484 | |
376.9 | |
Powder |
159°C to 161°C | |
C19H21ClN2O2S | |
10 mg | |
Soluble in ethanol or DMSO | |
C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O | |
376.899 | |
CHEBI:70773 | |
≥99% | |
Capsazepine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only