missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbadox, 98+%, Thermo Scientific Chemicals

$189.92 - $472.99

Chemical Identifiers

CAS 6804-07-5
MDL Number MFCD00057293
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6028414
View Documents
Thermo Scientific Chemicals
J6028414
25 g
Each for $189.92
 
AAJ6028422
View Documents
Thermo Scientific Chemicals
J6028422
100 g
Each for $472.99
 
Description

Description

Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

Solubility
Soluble in 1N NaOH (50 mg/ml), and water (partly).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

6804-07-5
MFCD00057293

Specifications

6804-07-5
Yellow
C13H10BrNO
25 g
14,1780
Soluble in 1N NaOH (50mg/ml),and water (partly).
C1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Br
276.13
262.22
Solid
239°C to 240°C
18°C (64°F)
MFCD00057293
UN1325
4-bromobenzanilide, n-phenyl 4-bromobenzamide, benzamide, 4-bromo-n-phenyl, 4-bromo-n-phenyl-benzamide, 4-bromo-n-phenyl-menzamide, maybridge1_004278, n1-phenyl-4-bromobenzamide, 4-bromophenyl-n-benzamide
QVTGXXQAVWWJRO-UHFFFAOYSA-N
4-bromo-N-phenylbenzamide
690966
≥98%
Carbadox
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H350-H302
Flammable solid.
May cause cancer.
Harmful if swallowed.

P201-P202-P210-P240-P241-P264b-P270-P281-P301+P312-P308+P313-P330-P370+P378q-P501c

H228-H302-H350

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : FE2779000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only