missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbenicillin disodium salt, Thermo Scientific Chemicals

Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities.

Supplier:  Thermo Scientific Chemicals J6194906

 View more versions of this product

Catalog No. AAJ6194906


Explore available promotions

Description

Description

Carbenicillin disodium salt is a water-soluble antibiotic which is effective against gram-negative bacteria.Carbenicillin disodium salt is widely utilized as an antibiotic which is used to control bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. It has been involved in the study of the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
  • In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium

Application

  • Due to its antibacterial activity, this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
  • It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
Specifications

Specifications

Carbenicillin Disodium Salt
Powder
White
C17H16N2Na2O6S
5722128
Soluble in water and alcohol.
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
422.36
CHEBI:34609
Carbenicillin disodium salt
4800-94-6
5 g
MFCD00077683
14,1792
RTYJTGSCYUUYAL-YCAHSCEMSA-L
disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
20933
422.36

Chemical Identifiers

4800-94-6
422.36
RTYJTGSCYUUYAL-YCAHSCEMSA-L
CHEBI:34609
C17H16N2Na2O6S
MFCD00077683
20933
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P271-P272-P280g-P285-P302+P352-P304+P340-P333+P313-P342+P311-P363-P501c

H317-H334-H335

EINECSNumber : 225-360-8

RTECSNumber : ON9105000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only